Assay Details
| Gene Symbol: Actr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106364.1 | NP_001099834.1 | 1 - 2 | 112 | 63 | |
| XM_006231470.2 | XP_006231532.1 | 1 - 2 | 224 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Actr1a
Genomic Map