Assay Details
| Gene Symbol: LOC100911951 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008760482.1 | XP_008758704.1 | 1 - 2 | 348 | 69 | |
| Gene Symbol: Kcnip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033961.1 | NP_001029133.1 | 1 - 2 | 293 | 69 | |
| GenBank mRNA | AF269285.1 | - | 1 - 2 | 71 | 69 | |
| BC085905.1 | - | 1 - 2 | 293 | 7193750 | 69 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100911951
Genomic Map