Assay Details
| Gene Symbol: Hlx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077674.1 | NP_001071142.1 | 2 - 3 | 1081 | 81 | |
| GenBank mRNA | BC127513.1 | - | 2 - 3 | 1081 | 7099599 | 81 |
More Information
Updated Mapping Information
Set Membership: |
|

Hlx
Genomic Map