Assay Details
| Gene Symbol: LOC317456 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047894.1 | NP_001041359.1 | 2 | 418 | 76 | |
| GenBank mRNA | AY383668.1 | - | 2 | 418 | 76 | |
| Gene Symbol: Lpgat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109376.2 | NP_001102846.1 | 4 - 5 | 817 | 76 | |
| XM_008769875.1 | XP_008768097.1 | 5 - 6 | 875 | 76 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC317456
Genomic Map