Assay Details
| Gene Symbol: Mpz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017027.1 | NP_058723.1 | 4 - 5 | 615 | 116 | |
| GenBank mRNA | BC078859.1 | - | 615 | 116 | ||
| K03242.1 | - | 4 - 5 | 615 | 116 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mpz
Genomic Map