Assay Details
| Gene Symbol: Ufc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003709.2 | NP_001003709.1 | 1 - 2 | 335 | 102 | |
| XM_006250251.2 | XP_006250313.1 | 2 - 3 | 290 | 102 | ||
| GenBank mRNA | AB186051.1 | - | 1 - 2 | 253 | 102 | |
| AB441169.1 | - | 1 - 2 | 253 | 102 | ||
| BC087648.1 | - | 1 - 2 | 335 | 7310651 | 102 | |
| FQ212748.1 | - | 1 - 2 | 353 | 102 | ||
| FQ213023.1 | - | 1 - 2 | 326 | 102 | ||
| FQ213507.1 | - | 1 - 2 | 336 | 102 | ||
| FQ216057.1 | - | 1 - 2 | 330 | 102 | ||
| FQ218926.1 | - | 1 - 2 | 356 | 102 | ||
| FQ232457.1 | - | 1 - 2 | 312 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ufc1
Genomic Map