Assay Details
| Gene Symbol: Dtl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006250494.2 | XP_006250556.1 | 12 - 13 | 1133 | 74 | |
| XM_223074.7 | XP_223074.5 | 12 - 13 | 1292 | 74 | ||
More Information
Set Membership: |
|

Dtl
Genomic Map