Assay Details
| Gene Symbol: LOC100911356 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751804.3 | XP_003751852.1 | 3 - 4 | 363 | 102 | |
| XM_008772208.1 | XP_008770430.1 | 3 - 4 | 363 | 102 | ||
| Gene Symbol: LOC103694912 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773908.1 | XP_008772130.1 | 2 - 3 | 607 | 102 | |
| Gene Symbol: Pex19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107375.2 | NP_001100845.1 | 3 - 4 | 370 | 102 | |
| NM_001134777.1 | NP_001128249.1 | 3 - 4 | 370 | 102 | ||
| GenBank mRNA | CB718412.1 | - | 3 - 4 | 383 | 102 | |
| CK600534.1 | - | 3 - 4 | 364 | 102 | ||
| Y09049.1 | - | 3 - 4 | 349 | 102 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100911356
Genomic Map