Assay Details
| Gene Symbol: Cr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005265.1 | NP_001005265.1 | 9 - 10 | 1522 | 132 | |
| NM_001005330.1 | NP_001005330.1 | 1894 | 132 | |||
| NM_019301.2 | NP_062174.1 | 10 - 11 | 1708 | 132 | ||
| GenBank mRNA | BC061736.1 | - | 10 - 11 | 1477 | 5599318 | 132 |
| D42114.1 | - | 9 - 10 | 1331 | 132 | ||
| D42115.1 | - | 10 - 11 | 1453 | 132 | ||
| FQ220559.1 | - | 6 - 7 | 727 | 132 | ||
| L36532.1 | - | 10 - 11 | 1476 | 132 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cr1l
Genomic Map