Assay Details
| Gene Symbol: Ly9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191673.1 | NP_001178602.1 | 7 - 8 | 1495 | 74 | |
| XM_008769732.1 | XP_008767954.1 | 7 - 8 | 1581 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ly9
Genomic Map