Assay Details
| Gene Symbol: LOC103691995 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008762593.1 | XP_008760815.1 | 2 - 3 | 352 | 112 | |
| Gene Symbol: Mrgbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173739.1 | NP_001167210.1 | 294 | 112 | ||
| GenBank mRNA | CO403993.1 | - | 294 | 112 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC103691995
Genomic Map