Assay Details
| Gene Symbol: Ogfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053340.2 | NP_445792.2 | 5 - 6 | 468 | 87 | |
| GenBank mRNA | BC101898.1 | - | 5 - 6 | 449 | 7932932 | 87 |
| EV778659.1 | - | 5 - 6 | 87 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Ogfr
Genomic Map