Assay Details
| Gene Symbol: Pcmtd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107810.1 | NP_001101280.2 | 4 - 5 | 695 | 78 | |
| GenBank mRNA | FQ211587.1 | - | 4 - 5 | 694 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pcmtd2
Genomic Map