Assay Details
| Gene Symbol: Zfp512b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107809.1 | NP_001101279.1 | 2 - 3 | 440 | 73 | |
| XM_008762511.1 | XP_008760733.1 | 2 - 3 | 481 | 73 | ||
| XM_008762512.1 | XP_008760734.1 | 2 - 3 | 241 | 73 | ||
| XM_008762513.1 | XP_008760735.1 | 2 - 3 | 241 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp512b
Genomic Map