Assay Details
| Gene Symbol: Lsm14b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002729258.4 | XP_002729304.3 | 2 - 3 | 488 | 146 | |
| XM_006235827.2 | XP_006235889.1 | 2 - 3 | 488 | 146 | ||
| XM_006235828.2 | XP_006235890.1 | 2 - 3 | 489 | 146 | ||
More Information
Set Membership: |
|

Lsm14b
Genomic Map