Assay Details
| Gene Symbol: Ocm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012995.1 | NP_037127.1 | 1 - 2 | 77 | 82 | |
| GenBank mRNA | J02705.1 | - | 1 - 2 | 77 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the J02705.1, NM_012995.1 transcripts.
Set Membership: |
|

Ocm2
Genomic Map