Assay Details
| Gene Symbol: Ocm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012995.1 | NP_037127.1 | 2 - 3 | 129 | 107 | |
| XM_006248852.2 | XP_006248914.1 | 2 - 3 | 311 | 107 | ||
| GenBank mRNA | J02705.1 | - | 2 - 3 | 129 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ocm2
Genomic Map