Assay Details
| Gene Symbol: Lnx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108329.1 | NP_001101799.1 | 9 - 10 | 2118 | 66 | |
| XM_006248835.2 | XP_006248897.1 | 9 - 10 | 2245 | 66 | ||
| XM_008768976.1 | XP_008767198.1 | 8 - 9 | 7164 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P21749 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P669103 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117006 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527896 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950725 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473707 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510871 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662877 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289977 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451716 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195663 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111364 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295371 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619668 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859574 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41899 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848668 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688452 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704714 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76088 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491867 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10319 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655708 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674500 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397582 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888639 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509915 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724049 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225624 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586301 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890882 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869080 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101499 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437080 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386283 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883306 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521806 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67423 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156364 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175165 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40806 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19220 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100016 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P669103 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P21749 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P669103 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P21749 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P669103 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P21749 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P21749 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 74823 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 67272 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10760073 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10760075 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10760074 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10760077 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10760076 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10760072 | - | - |
| Rat Genome 230 2.0 Array | 1383359_at | - | - |
| Rat Genome 230 2.0 Array | 1383359_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1200001162.rn.1 | TC1200000164.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200001163.rn.1 | TC1200000164.rn.1 | RTA TC |
| Rat Expression Set 230 | 1383359_PM_at | - | - |
| Rat Expression Set 230 | 1383359_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10760072 | - | - |
| Rat Genome U34 Set | rc_AI072958_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20798396 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1354119 | - | - |

Lnx2
Genomic Map