Assay Details
| Gene Symbol: Kdelr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013122.1 | NP_001013140.1 | 1 - 2 | 187 | 68 | |
| GenBank mRNA | BC085786.1 | - | 1 - 2 | 187 | 7108608 | 68 |
More Information
Updated Mapping Information
Set Membership: |
|

Kdelr2
Genomic Map