Assay Details
| Gene Symbol: Ppat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057198.2 | NP_476546.1 | 6 - 7 | 1294 | 66 | |
| GenBank mRNA | BC086999.1 | - | 6 - 7 | 1294 | 7189259 | 66 |
| D10853.1 | - | 6 - 7 | 802 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppat
Genomic Map