Assay Details
| Gene Symbol: Chic2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105736.2 | NP_001099206.1 | 2 - 3 | 223 | 112 | |
| GenBank mRNA | BE109715.1 | - | 3 - 4 | 201 | 112 | |
| EU202679.1 | - | 2 - 3 | 169 | 112 | ||
| EU202680.1 | - | 2 - 3 | 169 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Chic2
Genomic Map