Assay Details
| Gene Symbol: Noa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006959.1 | NP_001006960.1 | 3 - 4 | 1604 | 91 | |
| XM_006250845.2 | XP_006250907.1 | 3 - 4 | 1919 | 91 | ||
| GenBank mRNA | BC083902.1 | - | 3 - 4 | 1604 | 7133605 | 91 |
More Information
Updated Mapping Information
Set Membership: |
|

Noa1
Genomic Map