Assay Details
| Gene Symbol: Klf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105742.1 | NP_001099212.1 | 2 - 3 | 293 | 120 | |
| XM_006250995.2 | XP_006251057.1 | 2 - 3 | 5690 | 120 | ||
| XM_008770134.1 | XP_008768356.1 | 2 - 3 | 207 | 120 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Klf3
Genomic Map