Assay Details
| Gene Symbol: Klf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006250995.2 | XP_006251057.1 | 1 - 2 | 5590 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006250995.2 transcripts.
Set Membership: |
|

Klf3
Genomic Map