Assay Details
| Gene Symbol: Gnrhr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031038.3 | NP_112300.2 | 1 - 2 | 603 | 67 | |
| GenBank mRNA | L07646.1 | - | 1 - 2 | 589 | 67 | |
| X76635.1 | - | 1 - 2 | 542 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gnrhr
Genomic Map