Assay Details
| Gene Symbol: Fbxl15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107603.1 | NP_001101073.1 | 1 - 2 | 343 | 92 | |
| GenBank mRNA | FQ235302.1 | - | 1 - 2 | 335 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001107603.1 transcripts.
Set Membership: |
|

Fbxl15
Genomic Map