Assay Details
| Gene Symbol: Cyp2c23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031839.3 | NP_114027.2 | 2 - 3 | 370 | 83 | |
| GenBank mRNA | BC078707.1 | - | 2 - 3 | 370 | 7110580 | 83 |
| U04733.1 | - | 2 - 3 | 351 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cyp2c23
Genomic Map