Assay Details
| Gene Symbol: Cyp2c23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031839.3 | NP_114027.2 | 5 - 6 | 854 | 78 | |
| GenBank mRNA | BC078707.1 | - | 5 - 6 | 854 | 7110580 | 78 |
| FQ209508.1 | - | 3 - 4 | 453 | 78 | ||
| FQ218882.1 | - | 2 - 3 | 203 | 78 | ||
| U04733.1 | - | 5 - 6 | 835 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cyp2c23
Genomic Map