Assay Details
Gene Symbol: Prol1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133513.1 | NP_598197.1 | 1 - 2 | 107 | 94 | |
GenBank mRNA | U03407.1 | - | 1 - 2 | 107 | 94 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_133513.1, U03407.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
