Assay Details
| Gene Symbol: Pnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106031.1 | NP_001099501.1 | 2 - 3 | 244 | 95 | |
| XM_006251857.2 | XP_006251919.1 | 2 - 3 | 274 | 95 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pnp
Genomic Map