Assay Details
| Gene Symbol: Pnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006251857.2 | XP_006251919.1 | 5 - 6 | 746 | 73 | |
| GenBank mRNA | FQ209687.1 | - | 2 - 3 | 270 | 73 | |
| FQ213258.1 | - | 2 - 3 | 270 | 73 | ||
| FQ214084.1 | - | 2 - 3 | 270 | 73 | ||
| FQ218440.1 | - | 2 - 3 | 270 | 73 | ||
| FQ219529.1 | - | 2 - 3 | 270 | 73 | ||
| FQ225420.1 | - | 2 - 3 | 270 | 73 | ||
| FQ225955.1 | - | 2 - 3 | 270 | 73 | ||
| FQ228749.1 | - | 2 - 3 | 270 | 73 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pnp
Genomic Map