Assay Details
| Gene Symbol: Wdhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107255.1 | NP_001100725.1 | 20 - 21 | 2898 | 58 | |
| XM_006251795.2 | XP_006251857.1 | 20 - 21 | 2906 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Wdhd1
Genomic Map