Assay Details
| Gene Symbol: Naa30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109099.1 | NP_001102569.1 | 2 - 3 | 900 | 77 | |
| XM_006251813.2 | XP_006251875.1 | 3 - 4 | 1687 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Naa30
Genomic Map