Assay Details
| Gene Symbol: Acbd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182843.1 | NP_878263.1 | 1 - 2 | 328 | 103 | |
| GenBank mRNA | AY336075.1 | - | 1 - 2 | 328 | 103 | |
| BC083877.1 | - | 1 - 2 | 289 | 7130229 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acbd3
Genomic Map