Assay Details
| Gene Symbol: Dcaf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014231.1 | NP_001014253.1 | 3 - 4 | 281 | 77 | |
| XM_006250297.2 | XP_006250359.1 | 3 - 4 | 288 | 77 | ||
| XM_006250298.2 | XP_006250360.1 | 3 - 4 | 908 | 77 | ||
| XM_006250299.2 | XP_006250361.1 | 3 - 4 | 304 | 77 | ||
| XM_006250300.2 | XP_006250362.1 | 3 - 4 | 304 | 77 | ||
| XM_008769746.1 | XP_008767968.1 | 3 - 4 | 304 | 77 | ||
| GenBank mRNA | BC085957.1 | - | 3 - 4 | 281 | 7134976 | 77 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P396657 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391376 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179780 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426433 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967958 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725091 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213450 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333956 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35831 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745406 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432725 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763574 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534679 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252771 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187453 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347883 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403333 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268605 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341046 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634082 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34939 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259219 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 627971 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858991 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558437 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839090 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880817 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519875 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387693 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418983 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245258 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773028 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809390 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 627393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486217 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498457 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210380 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680164 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833651 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514944 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384216 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523148 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772573 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218289 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682516 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84327 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789726 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913681 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543891 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851453 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192734 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506597 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537409 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812569 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752524 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271953 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32078 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734925 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20651 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875946 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583908 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515802 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832850 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883122 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955054 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56002 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875495 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462890 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290694 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394928 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59115 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48315 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161342 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37508 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44214 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46399 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152784 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44718 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109102 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128226 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72826 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58157 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5419 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73952 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160212 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161452 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39904 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161948 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P396657 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P396657 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P396657 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P313918 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P457702 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 19283 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 50489 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 15968 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 50828 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 20335 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 45703 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 87501 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 74217 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 16800 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 24963 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 14882 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 58949 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 11854 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10765678 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708652 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10765674 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10765672 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10765681 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10765679 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10765677 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10765682 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10765673 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10765675 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10765676 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10765680 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10708651 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10765671 | - | - |
| Rat Genome 230 2.0 Array | 1399138_at | - | - |
| Rat Genome 230 2.0 Array | 1399138_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1399138_at | - | - |
| Affymetrix HT Rat Focus Array | 1399138_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1300005572.rn.1 | TC1300000780.rn.1 | RTA TC |
| Rat Expression Set 230 | 1399138_at | - | - |
| Rat Expression Set 230 | 1399138_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10765671 | - | - |
| Rat Genome U34 Set | rc_AI044727_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1650574 | - | - |
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1530374 | - | - |
| RatRef-12 Expression BeadChip | ILMN_161048 | - | - |

Dcaf8
Genomic Map