Assay Details
| Gene Symbol: Ovol2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106519.1 | NP_001099989.1 | 2 - 3 | 287 | 94 | |
| XM_006235124.2 | XP_006235186.1 | 2 - 3 | 1446 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ovol2
Genomic Map