Assay Details
| Gene Symbol: Dtd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108594.1 | NP_001102064.1 | 2 - 3 | 198 | 71 | |
| XM_006235128.2 | XP_006235190.1 | 2 - 3 | 238 | 71 | ||
| GenBank mRNA | BC159413.1 | - | 2 - 3 | 181 | 8367451 | 71 |
| FQ214547.1 | - | 2 - 3 | 170 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P410192 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288408 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352687 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306238 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285573 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418824 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70760 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656272 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106597 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778204 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375517 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157238 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159640 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118840 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585281 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940613 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22818 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832634 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884586 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819698 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660717 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403842 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289258 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820030 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422531 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660751 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949707 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790906 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327064 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495227 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504156 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475311 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41181 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456927 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243991 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492300 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729932 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154343 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655837 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558938 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578454 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272500 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697020 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633510 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267469 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692247 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295212 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317581 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526642 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278346 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232403 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541394 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529590 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59208 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279996 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385942 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174986 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63508 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549406 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839621 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643590 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107921 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337436 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484601 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17167 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222339 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220775 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802147 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101666 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611007 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445345 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763099 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828342 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178070 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277398 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733830 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17190 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147800 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915745 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110541 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483592 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491472 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512971 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292811 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219736 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734286 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130970 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 180579 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84221 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52641 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157615 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46277 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18826 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121613 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15840 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24731 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129044 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175642 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76758 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53697 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94627 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167070 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52117 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126253 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10885 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P410192 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P410192 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P410192 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P410192 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 29139 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 9399 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 97753 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 53719 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10840421 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10840423 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10840419 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10840424 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10840422 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10840420 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10840418 | - | - |
| Rat Genome 230 2.0 Array | 1388428_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1388428_at | - | - |
| Affymetrix HT Rat Focus Array | 1388428_at | - | - |
| Affymetrix HT Rat Focus Array | 1388428_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0300013559.rn.1 | TC0300001890.rn.1 | RTA TC |
| Rat Expression Set 230 | 1388428_PM_at | - | - |
| Rat Expression Set 230 | 1388428_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10840418 | - | - |
| Rat Genome U34 Set | rc_AI233365_at | - | - |
| Rat Genome U34 Set | rc_AI233365_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21574106 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1364969 | - | - |

Dtd1
Genomic Map