Assay Details
| Gene Symbol: Cst13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109343.1 | NP_001102813.1 | 2 - 3 | 457 | 87 | |
| XM_006235162.2 | XP_006235224.1 | 3 - 4 | 1048 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cst13
Genomic Map