Assay Details
| Gene Symbol: Kif16b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107783.2 | NP_001101253.2 | 7 - 8 | 755 | 64 | |
| XM_006235108.2 | XP_006235170.1 | 7 - 8 | 908 | 64 | ||
| XM_006235109.2 | XP_006235171.1 | 7 - 8 | 908 | 64 | ||
| XM_006235112.2 | XP_006235174.1 | 7 - 8 | 890 | 64 | ||
| XM_006235113.2 | XP_006235175.1 | 7 - 8 | 890 | 64 | ||
| XM_008762263.1 | XP_008760485.1 | 8 - 9 | 796 | 64 | ||
| XM_008762264.1 | XP_008760486.1 | 7 - 8 | 720 | 64 | ||
| XM_008762265.1 | XP_008760487.1 | 7 - 8 | 1484 | 64 | ||
| XM_008762266.1 | XP_008760488.1 | 6 - 7 | 793 | 64 | ||
| XM_008762267.1 | XP_008760489.1 | 7 - 8 | 890 | 64 | ||
| XM_008762268.1 | XP_008760490.1 | 7 - 8 | 890 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kif16b
Genomic Map