Assay Details
| Gene Symbol: Gid8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198743.1 | NP_942038.1 | 1 - 2 | 54 | 73 | |
| GenBank mRNA | AY156921.1 | - | 1 - 2 | 54 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AY156921.1, NM_198743.1 transcripts.
Set Membership: |
|

Gid8
Genomic Map