Assay Details
| Gene Symbol: Gid8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198743.1 | NP_942038.1 | 2 - 3 | 182 | 76 | |
| GenBank mRNA | AY156921.1 | - | 2 - 3 | 182 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gid8
Genomic Map