Assay Details
| Gene Symbol: Lime1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108614.1 | NP_001102084.1 | 2 - 3 | 1096 | 155 | |
| XM_008762521.1 | XP_008760743.1 | 3 - 4 | 672 | 155 | ||
| GenBank mRNA | BC158665.1 | - | 2 - 3 | 586 | 155 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lime1
Genomic Map