Assay Details
| Gene Symbol: Car5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005551.2 | NP_001005551.1 | 1 - 2 | 200 | 104 | |
| XM_006256859.2 | XP_006256921.1 | 2 - 3 | 297 | 104 | ||
| GenBank mRNA | BC081872.1 | - | 1 - 2 | 202 | 7106842 | 104 |
More Information
Updated Mapping Information
Set Membership: |
|

Car5b
Genomic Map