Assay Details
| Gene Symbol: Car5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005551.2 | NP_001005551.1 | 3 - 4 | 517 | 100 | |
| XM_006256859.2 | XP_006256921.1 | 4 - 5 | 614 | 100 | ||
| GenBank mRNA | BC081872.1 | - | 3 - 4 | 519 | 7106842 | 100 |
More Information
Updated Mapping Information
Set Membership: |
|

Car5b
Genomic Map