Assay Details
| Gene Symbol: Figf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031761.1 | NP_113949.1 | 2 - 3 | 591 | 85 | |
| XM_006256863.2 | XP_006256925.1 | 3 - 4 | 738 | 85 | ||
| XM_006256864.2 | XP_006256926.1 | 2 - 3 | 1068 | 85 | ||
| GenBank mRNA | AF014827.1 | - | 2 - 3 | 591 | 85 | |
| AY032728.1 | - | 2 - 3 | 366 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Figf
Genomic Map