Assay Details
| Gene Symbol: Figf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006256863.2 | XP_006256925.1 | 5 - 6 | 1070 | 83 | |
| XM_006256864.2 | XP_006256926.1 | 4 - 5 | 1400 | 83 | ||
| GenBank mRNA | AY032728.1 | - | 4 - 5 | 698 | 83 | |
More Information
Set Membership: |
|

Figf
Genomic Map