Assay Details
| Gene Symbol: Zrsr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006256887.2 | XP_006256949.1 | 7 - 8 | 694 | 169 | |
| XM_217612.5 | XP_217612.5 | 6 - 7 | 817 | 169 | ||
More Information
Set Membership: |
|

Zrsr2
Genomic Map