Assay Details
| Gene Symbol: LOC102550809 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006256902.2 | XP_006256964.1 | 5 - 6 | 2033 | 79 | |
| XM_008773212.1 | XP_008771434.1 | 4 - 5 | 3491 | 79 | ||
More Information
Set Membership: |
|

LOC102550809
Genomic Map