Assay Details
| Gene Symbol: Cyb5rl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001069267.4 | XP_001069267.2 | 2 - 3 | 358 | 123 | |
| XM_006238551.2 | XP_006238613.1 | 2 - 3 | 360 | 123 | ||
| XM_008763922.1 | XP_008762144.1 | 2 - 3 | 347 | 123 | ||
| XM_008763923.1 | XP_008762145.1 | 2 - 3 | 348 | 123 | ||
| XM_008763924.1 | XP_008762146.1 | 2 - 3 | 346 | 123 | ||
| XM_008763925.1 | XP_008762147.1 | 2 - 3 | 334 | 123 | ||
| XM_008763926.1 | XP_008762148.1 | 2 - 3 | 338 | 123 | ||
| XM_008763927.1 | XP_008762149.1 | 2 - 3 | 342 | 123 | ||
| XM_008763928.1 | XP_008762150.1 | 2 - 3 | 359 | 123 | ||
| XM_008763929.1 | XP_008762151.1 | 2 - 3 | 359 | 123 | ||
| XM_008763930.1 | XP_008762152.1 | 2 - 3 | 458 | 123 | ||
More Information
Set Membership: |
|

Cyb5rl
Genomic Map