Assay Details
| Gene Symbol: Cyb5rl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001069267.4 | XP_001069267.2 | 4 - 5 | 596 | 74 | |
| XM_006238551.2 | XP_006238613.1 | 4 - 5 | 598 | 74 | ||
| XM_008763922.1 | XP_008762144.1 | 4 - 5 | 585 | 74 | ||
| XM_008763923.1 | XP_008762145.1 | 4 - 5 | 586 | 74 | ||
| XM_008763924.1 | XP_008762146.1 | 4 - 5 | 584 | 74 | ||
| XM_008763925.1 | XP_008762147.1 | 4 - 5 | 572 | 74 | ||
| XM_008763926.1 | XP_008762148.1 | 4 - 5 | 576 | 74 | ||
| XM_008763927.1 | XP_008762149.1 | 4 - 5 | 580 | 74 | ||
| XM_008763930.1 | XP_008762152.1 | 4 - 5 | 696 | 74 | ||
More Information
Set Membership: |
|

Cyb5rl
Genomic Map